accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101209-08-9

IUPAC Name: N-(4-acetyl-2-bromophenyl)acetamide
Synonyms: N-(4-acetyl-2-bromophenyl)acetamide
Molecular Weight (g/mol): 256.1
Molecular Formula: C10H10BrNO2
InChi Key: PMYJAVHDFDKJBS-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(Br)C=C(C=C1)C(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results