CAS RN 101224-22-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cerilliant™ Buprenorphine Glucuronide Solution, 100 μg/mL in methanol, Certified Reference Material, MilliporeSigma™ Supelco™
Buprenorphine glucuronide is a major urinary metabolite of the semi-synthetic opioid, buprenorphine.