accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaNaNaNaOPOOPOOOPOOOPOOOPOOOPOOO

CAS RN 10124-56-8

IUPAC Name: hexasodium hexaoxocyclohexaphosphoxane-2,4,6,8,10,12-hexakis(olate)
Synonyms: sodium hexametaphosphate
Molecular Weight (g/mol): 611.76
Molecular Formula: Na6O18P6
InChi Key: GCLGEJMYGQKIIW-UHFFFAOYSA-H
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P1(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)OP([O-])(=O)O1
Grade 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)