accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101251-09-6

IUPAC Name: (4-acetamidophenyl)boronic acid
Synonyms: 4-acetamidophenylboronic acid
Molecular Weight (g/mol): 178.98
Molecular Formula: C8H10BNO3
InChi Key: VYEWTHXZHHATTA-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)B(O)O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results