accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOO

CAS RN 10128-71-9

IUPAC Name: 3-hydroxypyridine-4-carboxylate
Synonyms: 3-hydroxypyridine-4-carboxylate
Molecular Weight (g/mol): 138.1
Molecular Formula: C6H4NO3
InChi Key: JEHGATQUCUYHJL-UHFFFAOYSA-M
SMILES: OC1=CN=CC=C1C([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)