accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10128-91-3

IUPAC Name: methyl 2-oxo-1,2-dihydropyridine-3-carboxylate
Synonyms: methyl 2-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: SILBTMNCGYLTOK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CNC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results