accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOOCl

CAS RN 10130-74-2

IUPAC Name: 3-methoxybenzene-1-sulfonyl chloride
Synonyms: 3-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 206.64
Molecular Formula: C7H7ClO3S
InChi Key: JHJKSEKUZNJKGO-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)