accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNNFFFSOONHSOOFFF

CAS RN 1013027-27-4

IUPAC Name: 1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide; 1-ethenyl-1H-imidazole
Synonyms: 1,1,1-trifluoro-N-trifluoromethanesulfonylmethanesulfonamide; 1-ethenylimidazole
Molecular Weight (g/mol): 375.26
Molecular Formula: C7H7F6N3O4S2
InChi Key: OVFZHDDKAVGFIS-UHFFFAOYSA-N
SMILES: C=CN1C=CN=C1.FC(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)