accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10132-01-1

IUPAC Name: 1-chloro-4-phenylphthalazine
Synonyms: 1-chloro-4-phenylphthalazine
Molecular Weight (g/mol): 240.69
Molecular Formula: C14H9ClN2
InChi Key: WJJDLSHYLZRFDD-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=C(N=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results