accessibility menu, dialog, popup

UserName

Viewing profile as user:
FICl

CAS RN 101335-11-9

IUPAC Name: 2-chloro-4-fluoro-1-iodobenzene
Synonyms: 2-chloro-4-fluoro-1-iodobenzene
Molecular Weight (g/mol): 256.44
Molecular Formula: C6H3ClFI
InChi Key: POTCKVPDYXEGSV-UHFFFAOYSA-N
SMILES: FC1=CC=C(I)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)