accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNHCH3NSH3CN

CAS RN 1014-70-6

IUPAC Name: N2,N4-diethyl-6-(methylsulfanyl)-1,3,5-triazine-2,4-diamine
Synonyms: simetryn gy-bon
Molecular Weight (g/mol): 213.3
Molecular Formula: C8H15N5S
InChi Key: MGLWZSOBALDPEK-UHFFFAOYSA-N
SMILES: CCNC1=NC(SC)=NC(NCC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results