accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOFFF

CAS RN 1014-81-9

IUPAC Name: 3-(trifluoromethoxy)benzoic acid
Synonyms: 3-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: OKPFIWIMBJNFSE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(OC(F)(F)F)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results