CAS RN 1014-93-3
CAS RN 1014-93-3
IUPAC Name:
{[(5-bromopentyl)oxy]methyl}benzene
Synonyms:
{[(5-bromopentyl)oxy]methyl}benzene
Molecular Weight (g/mol):
257.17
Molecular Formula:
C12H17BrO
InChi Key:
HGUZJKJSYSSVLK-UHFFFAOYSA-N
SMILES:
BrCCCCCOCC1=CC=CC=C1
1
–
1
of
1
results
1
Benzyl 5-Bromoamyl Ether 95.0+%, TCI America™

CAS: 1014-93-3 Molecular Formula: C12H17BrO Molecular Weight (g/mol): 257.17 MDL Number: MFCD02258456 InChI Key: HGUZJKJSYSSVLK-UHFFFAOYSA-N Synonym: Benzyl 5-Bromopentyl Ether, Benzyloxypentyl Bromide, (5-Bromopentyloxymethyl)benzene PubChem CID: 14221121 IUPAC Name: {[(5-bromopentyl)oxy]methyl}benzene SMILES: BrCCCCCOCC1=CC=CC=C1