accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101420-81-9

IUPAC Name: 3-bromo-4-nitrobenzoic acid
Synonyms: 3-bromo-4-nitrobenzoic acid
Molecular Weight (g/mol): 246.02
Molecular Formula: C7H4BrNO4
InChi Key: KKPPNEJUUOQRLE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)