accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101426-31-7

H3COONSCH3

CAS RN 101426-31-7

IUPAC Name: 2-(4-methyl-1,3-thiazol-5-yl)ethyl decanoate
Synonyms: 2-(4-methyl-1,3-thiazol-5-yl)ethyl decanoate
Molecular Weight (g/mol): 297.46
Molecular Formula: C16H27NO2S
InChi Key: RUOLKIITSCGKJQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(=O)OCCC1=C(C)N=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results