accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10147-36-1

IUPAC Name: propane-1-sulfonyl chloride
Synonyms: propane-1-sulfonyl chloride
Molecular Weight (g/mol): 142.6
Molecular Formula: C3H7ClO2S
InChi Key: KPBSJEBFALFJTO-UHFFFAOYSA-N
SMILES: CCCS(Cl)(=O)=O
Boiling Point 
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)