accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1015-38-9

IUPAC Name: diphenyl(sulfanyl)-λ⁵-phosphanethione
Synonyms: diphenyl(sulfanyl)-λ⁵-phosphanethione
Molecular Weight (g/mol): 250.31
Molecular Formula: C12H11PS2
InChi Key: CLUOCCWZZAGLPM-UHFFFAOYSA-N
SMILES: SP(=S)(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)