accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFFClF

CAS RN 101513-77-3

IUPAC Name: 3-chloro-2,4,5-trifluorobenzoate
Synonyms: 3-chloro-2,4,5-trifluorobenzoate
Molecular Weight (g/mol): 209.53
Molecular Formula: C7HClF3O2
InChi Key: KBESHLYCSZINAJ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=C(F)C(Cl)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results