accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOBrBr

CAS RN 1015423-45-6

IUPAC Name: dibromo-1H,3H-thieno[3,4-c]furan-1,3-dione
Synonyms: dibromothieno[3,4-c]furan-1,3-dione
Molecular Weight (g/mol): 311.93
Molecular Formula: C6Br2O3S
InChi Key: OQMJQMGTDSIYNQ-UHFFFAOYSA-N
SMILES: BrC1=C2C(=O)OC(=O)C2=C(Br)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results