accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1015691-13-0

IUPAC Name: tetraethylazanium hydrate chloride
Synonyms: tetraethylammonium hydrate chloride
Molecular Weight (g/mol): 183.72
Molecular Formula: C8H22ClNO
InChi Key: CALLTGJPWMIDPC-UHFFFAOYSA-M
SMILES: O.[Cl-].CC[N+](CC)(CC)CC
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results