accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1015937-56-0

IUPAC Name: potassium 2-hydroxy-3,5-diiodobenzoate
Synonyms: potassium 2-hydroxy-3,5-diiodobenzoate
Molecular Weight (g/mol): 428.01
Molecular Formula: C7H3I2KO3
InChi Key: SNHIVWOGVZUVLO-UHFFFAOYSA-M
SMILES: [K+].OC1=C(I)C=C(I)C=C1C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results