accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSO

CAS RN 1016-05-3

IUPAC Name: 8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
Synonyms: 8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
Molecular Weight (g/mol): 216.25
Molecular Formula: C12H8O2S
InChi Key: IKJFYINYNJYDTA-UHFFFAOYSA-N
SMILES: O=S1(=O)C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)