accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1016-77-9

IUPAC Name: (3-bromophenyl)(phenyl)methanone
Synonyms: 3-bromophenyl phenyl ketone
Molecular Weight (g/mol): 261.12
Molecular Formula: C13H9BrO
InChi Key: XNUMUNIJQMSNNN-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)