CAS RN 10163-15-2
IUPAC Name:
disodium phosphorofluoridate
Synonyms:
sodium monofluorophosphate
Molecular Weight (g/mol):
143.95
Molecular Formula:
FNa2O3P
InChi Key:
BFDWBSRJQZPEEB-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]P([O-])(F)=O
1
–
1
of
1
results
1
Sigma Aldrich Sodium fluorophosphate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.