CAS RN 1016748-73-4
IUPAC Name:
N-benzyl-4-chloro-N-(propan-2-yl)butanamide
Synonyms:
N-benzyl-4-chloro-N-isopropylbutanamide
Molecular Weight (g/mol):
253.77
Molecular Formula:
C14H20ClNO
InChi Key:
BEGDLNHPGOYQHE-UHFFFAOYSA-N
SMILES:
CC(C)N(CC1=CC=CC=C1)C(=O)CCCCl
1
–
1
of
1
results
1
N-Benzyl-4-chloro-N-isopropylbutyramide, 97%, Thermo Scientific™
CAS: 1016748-73-4 Molecular Formula: C14H20ClNO Molecular Weight (g/mol): 253.77 MDL Number: MFCD09807999 InChI Key: BEGDLNHPGOYQHE-UHFFFAOYSA-N Synonym: n-benzyl-4-chloro-n-isopropylbutanamide,n-benzyl-4-chloro-n-isopropylbutyramide,n-benzyl-4-chloro-n-propan-2-yl butanamide PubChem CID: 20119644 IUPAC Name: N-benzyl-4-chloro-N-propan-2-ylbutanamide SMILES: CC(C)N(CC1=CC=CC=C1)C(=O)CCCCl