accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10169-55-8

IUPAC Name: 1-[4-(phenylsulfanyl)phenyl]ethan-1-one
Synonyms: 1-[4-(phenylsulfanyl)phenyl]ethanone
Molecular Weight (g/mol): 228.31
Molecular Formula: C14H12OS
InChi Key: XUDYHODVSUXRPW-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(SC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results