accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10170-68-0

IUPAC Name: tris(oxolane); trichlorochromium
Synonyms: tris(THF); chromium chloride
Molecular Weight (g/mol): 374.67
Molecular Formula: C12H24Cl3CrO3
InChi Key: CYOMBOLDXZUMBU-UHFFFAOYSA-K
SMILES: Cl[Cr](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results