accessibility menu, dialog, popup

UserName

Viewing profile as user:
MnMnCOCOCOCOCOCOCOCOCOCO

CAS RN 10170-69-1

IUPAC Name: decakis(methanidylidyneoxidanium) dimanganese
Synonyms: dimanganese decacarbonyl
Molecular Weight (g/mol): 389.98
Molecular Formula: C10Mn2O10
InChi Key: QFEOTYVTTQCYAZ-UHFFFAOYSA-N
SMILES: [Mn].[Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results