accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10171-92-3

IUPAC Name: 1,5-dimethyl 2-methylpentanedioate
Synonyms: dimethyl methylglutarate
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: ZWKKRUNHAVNSFW-UHFFFAOYNA-N
SMILES: COC(=O)CCC(C)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results