accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101772-29-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-hydroxypropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)(methyl)amino]-3-hydroxypropanoic acid
Molecular Weight (g/mol): 219.24
Molecular Formula: C9H17NO5
InChi Key: NJQGIDVCNBZXLG-UHFFFAOYNA-N
SMILES: CN(C(CO)C(O)=O)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)