accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1017778-64-1

IUPAC Name: 3,4-difluoro-5-methylbenzaldehyde
Synonyms: 3,4-difluoro-5-methylbenzaldehyde
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: JYJHDXILRBVZBI-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results