accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101819-99-2

IUPAC Name: 6-amino-2,4-dichloro-3-ethylphenol hydrochloride
Synonyms: 6-amino-2,4-dichloro-3-ethylphenol hydrochloride
Molecular Weight (g/mol): 242.52
Molecular Formula: C8H10Cl3NO
InChi Key: XZZITYVICUAZNB-UHFFFAOYSA-N
SMILES: Cl.CCC1=C(Cl)C=C(N)C(O)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results