accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 101854-42-6

IUPAC Name: 4-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}butanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 4-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]butanoic acid; dicha
Molecular Weight (g/mol): 533.71
Molecular Formula: C29H47N3O6
InChi Key: CBSVEVKFQHTZSP-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(CCNC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results