accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1019-52-9

IUPAC Name: 4-hydroxy-3,5-dinitrobenzoic acid
Synonyms: 4-hydroxy-3,5-dinitrobenzoic acid
Molecular Weight (g/mol): 228.12
Molecular Formula: C7H4N2O7
InChi Key: GBSWIDSKAJFWMF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results