accessibility menu, dialog, popup

UserName

Viewing profile as user:
HSOOOOSH

CAS RN 10193-95-0

IUPAC Name: 4-[(2-sulfanylacetyl)oxy]butyl 2-sulfanylacetate
Synonyms: 4-[(2-sulfanylacetyl)oxy]butyl 2-sulfanylacetate
Molecular Weight (g/mol): 238.32
Molecular Formula: C8H14O4S2
InChi Key: IPNDIMIIGZSERC-UHFFFAOYSA-N
SMILES: SCC(=O)OCCCCOC(=O)CS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results