accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONOOOZnH2OH2OH2OH2OH2OH2O

CAS RN 10196-18-6

IUPAC Name: zinc(2+) hexahydrate dinitrate
Synonyms: zinc(2+) hexahydrate dinitrate
Molecular Weight (g/mol): 297.48
Molecular Formula: H12N2O12Zn
InChi Key: JGPSMWXKRPZZRG-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (18)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
Grade 
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (8)
Physical Form 
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (6)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (6)
  • (8)
  • (3)
  • (2)
  • (6)
  • (18)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (8)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)