accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COHONHONHCH3CH3H3C

CAS RN 101968-85-8

IUPAC Name: 2-[(tert-butylcarbamoyl)amino]-3,3-dimethylbutanoic acid
Synonyms: 2-[(tert-butylcarbamoyl)amino]-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 230.31
Molecular Formula: C11H22N2O3
InChi Key: RAAPXVRHYBAJQU-UHFFFAOYNA-N
SMILES: CC(C)(C)NC(=O)NC(C(O)=O)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)