accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHF

CAS RN 101969-75-9

IUPAC Name: 4-(2-fluorobenzoyl)phenol
Synonyms: 4-(2-fluorobenzoyl)phenol
Molecular Weight (g/mol): 216.21
Molecular Formula: C13H9FO2
InChi Key: FHFDJJWFCDGBEM-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results