accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFNNHO

CAS RN 101975-10-4

IUPAC Name: 6-[4-(difluoromethoxy)-3-methoxyphenyl]-2,3-dihydropyridazin-3-one
Synonyms: zardaverine zardaverina zardaverinum
Molecular Weight (g/mol): 268.22
Molecular Formula: C12H10F2N2O3
InChi Key: HJMQDJPMQIHLPB-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1OC(F)F)C1=NNC(=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results