accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10199-00-5

NNNN

CAS RN 10199-00-5

IUPAC Name: 2,2'-bipyrazine
Synonyms: 2,2'-bipyrazine
Molecular Weight (g/mol): 158.16
Molecular Formula: C8H6N4
InChi Key: DFXNVSIALRDJHY-UHFFFAOYSA-N
SMILES: C1=CN=C(C=N1)C1=NC=CN=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)