accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH2N

CAS RN 10199-50-5

IUPAC Name: 1-methyl-3-phenyl-1H-pyrazol-5-amine
Synonyms: 2-methyl-5-phenylpyrazol-3-amine
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: KCYRMURRLLYLPU-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1N)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)