accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3S

CAS RN 1019983-99-3

IUPAC Name: 7,17-dimethyl-6,16-dithiapentacyclo[11.7.0.0³,¹¹.0⁵,⁹.0¹⁵,¹⁹]icosa-1,3,5(9),7,10,12,14,17,19-nonaene
Synonyms: 7,17-dimethyl-6,16-dithiapentacyclo[11.7.0.0³,¹¹.0⁵,⁹.0¹⁵,¹⁹]icosa-1,3,5(9),7,10,12,14,17,19-nonaene
Molecular Weight (g/mol): 318.45
Molecular Formula: C20H14S2
InChi Key: HPJACFAPBXKLQA-UHFFFAOYSA-N
SMILES: CC1=CC2=CC3=CC4=CC5=C(C=C(C)S5)C=C4C=C3C=C2S1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results