accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-01-2

IUPAC Name: 3-oxo-N-phenylbutanamide
Synonyms: acetoacetanilide
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: DYRDKSSFIWVSNM-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)