accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-06-7

IUPAC Name: N,N''-diphenylguanidine
Synonyms: 1,3-diphenylguanidine
Molecular Weight (g/mol): 211.27
Molecular Formula: C13H13N3
InChi Key: OWRCNXZUPFZXOS-UHFFFAOYSA-N
SMILES: NC(NC1=CC=CC=C1)=NC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)