accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-09-0

IUPAC Name: diphenyl carbonate
Synonyms: diphenyl carbonate Ph2CO3 phenyl carbonate ((pho)2CO) (PhO)2CO
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: ROORDVPLFPIABK-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=C1)OC1=CC=CC=C1
Boiling Point 
  • (3)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (3)
  • (4)
  • (2)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)