accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHH3C

CAS RN 102-27-2

IUPAC Name: N-ethyl-3-methylaniline
Synonyms: benzenamine, N-ethyl-3-methyl-
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: GUYMMHOQXYZMJQ-UHFFFAOYSA-N
SMILES: CCNC1=CC=CC(C)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)