accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-36-3

IUPAC Name: 1,2-dichloro-4-isocyanatobenzene
Synonyms: 3,4-dichlorphenylisocyanate
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: MFUVCHZWGSJKEQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1Cl)N=C=O
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)