CAS RN 102-40-9
IUPAC Name:
2-[3-(2-hydroxyethoxy)phenoxy]ethan-1-ol
Synonyms:
2-[3-(2-hydroxyethoxy)phenoxy]ethanol
Molecular Weight (g/mol):
198.22
Molecular Formula:
C10H14O4
InChi Key:
IAXFZZHBFXRZMT-UHFFFAOYSA-N
SMILES:
OCCOC1=CC(OCCO)=CC=C1
1
–
1
of
1
results
1
1,3-Bis(2-hydroxyethoxy)benzene 98.0+%, TCI America™
CAS: 102-40-9 Molecular Formula: C10H14O4 Molecular Weight (g/mol): 198.22 MDL Number: MFCD00016566 InChI Key: IAXFZZHBFXRZMT-UHFFFAOYSA-N Synonym: Resorcinol Bis(beta-hydroxyethyl) Ether PubChem CID: 66885 IUPAC Name: 2-[3-(2-hydroxyethoxy)phenoxy]ethan-1-ol SMILES: OCCOC1=CC(OCCO)=CC=C1