accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CCH3

CAS RN 102-69-2

IUPAC Name: tripropylamine
Synonyms: tripropylamine NPr3
Molecular Weight (g/mol): 143.27
Molecular Formula: C9H21N
InChi Key: YFTHZRPMJXBUME-UHFFFAOYSA-N
SMILES: CCCN(CCC)CCC
Boiling Point 
  • (4)
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)