CAS RN 102-76-1
Triacetin, 99%
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.21 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N Synonym: triacetin,glyceryl triacetate,glycerol triacetate,glycerin triacetate,enzactin,triacetine,triacetylglycerol,fungacetin,glyped,triacetyl glycerine PubChem CID: 5541 ChEBI: CHEBI:9661 IUPAC Name: 2,3-diacetyloxypropyl acetate SMILES: CC(=O)OCC(COC(=O)C)OC(=O)C
Triacetin 99.0+%, TCI America™
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.205 MDL Number: MFCD00008716 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N Synonym: triacetin,glyceryl triacetate,glycerol triacetate,glycerin triacetate,enzactin,triacetine,triacetylglycerol,fungacetin,glyped,triacetyl glycerine PubChem CID: 5541 ChEBI: CHEBI:9661 IUPAC Name: 2,3-diacetyloxypropyl acetate SMILES: CC(=O)OCC(COC(=O)C)OC(=O)C
Triacetin, FCC, 98.5%, Spectrum™ Chemical
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.21 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N IUPAC Name: 1,3-bis(acetyloxy)propan-2-yl acetate SMILES: CC(=O)OCC(COC(C)=O)OC(C)=O
Triacetin, USP, 97-102%, Spectrum™ Chemical
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.21 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N IUPAC Name: 1,3-bis(acetyloxy)propan-2-yl acetate SMILES: CC(=O)OCC(COC(C)=O)OC(C)=O
Triacetin, USP, 97-102%, Spectrum™ Chemical
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.21 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N IUPAC Name: 1,3-bis(acetyloxy)propan-2-yl acetate SMILES: CC(=O)OCC(COC(C)=O)OC(C)=O
Triacetin, 99%, MP Biomedicals™
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.205 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N Synonym: triacetin,glyceryl triacetate,glycerol triacetate,glycerin triacetate,enzactin,triacetine,triacetylglycerol,fungacetin,glyped,triacetyl glycerine PubChem CID: 5541 ChEBI: CHEBI:9661 IUPAC Name: 2,3-diacetyloxypropyl acetate SMILES: CC(=O)OCC(COC(=O)C)OC(=O)C
Glycerol triacetate, 99%
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.205 MDL Number: MFCD00008716 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N Synonym: triacetin,glyceryl triacetate,glycerol triacetate,glycerin triacetate,enzactin,triacetine,triacetylglycerol,fungacetin,glyped,triacetyl glycerine PubChem CID: 5541 ChEBI: CHEBI:9661 IUPAC Name: 2,3-diacetyloxypropyl acetate SMILES: CC(=O)OCC(COC(=O)C)OC(=O)C
Triacetin (Glycerol Triacetate), Reagents
CAS: 102-76-1 Molecular Formula: C9H14O6 Molecular Weight (g/mol): 218.21 InChI Key: URAYPUMNDPQOKB-UHFFFAOYSA-N Synonym: 1,2,3-Propanetriol, Glycerol Triacetate IUPAC Name: 1,3-bis(acetyloxy)propan-2-yl acetate SMILES: CC(=O)OCC(COC(C)=O)OC(C)=O
Sigma Aldrich Triacetin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Triacetin, CP, Spectrum™ Chemical
CAS: 102-76-1